Howard Leight AirSoft® Earplugs, Honeywell Safety
Supplier: Honeywell Safety Products
DPAS-30R AS-30R
55533-014CS 3775.66 USD InStock
55533-014 55533-012 82001-017 82007-210
Howard Leight AirSoft® Earplugs, Honeywell Safety
Earplugs Reusable Earplugs
Reusable earplugs feature air pockets encased by shells of soft attenuating material, comfortable even for extended wear
  • Comfortable, even for extended wear
  • Washable, reusable protection
  • Available with or without connecting cord
Washable with soap and water for months of protection. Available with or without connecting cord. Color: blue.
Certificaciones: Meet ANSI S3.19 standard. CSA certified. CE marked.
Información para pedidos: Packaged in clear polybag of one pair each.
Order Now
ORDER
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
Collapse All
Description Noise reduction rating Supplier No. VWR Catalog Number Unit Price Quantity
Earplugs with red poly cord 27 dB DPAS-30R 55533-014 Pack of 100
Retrieving
Case of 1000
Retrieving
CSA Certified
Earplugs with red poly cord 27 dB AS-30R 82007-210 Case of 500
Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty