Protease Estimation Assay, G-Biosciences
Supplier: G-BIOSCIENCES
786-137
82021-296EA 436.76 USD InStock
82021-296
Protease Estimation Assay, G-Biosciences
Assays Protein Assays
The Protease Screening Kit provides you with a simple and quick method for testing your samples for proteolysis
Accessories: Resorufin-Casein Protease Substrate (#786-321): A colorimetric substrate that when treated with proteases releases resorufin that has an absorbance of 570nm. Supplied lyophilized. FITC-Casein Protease Substrate (#786-322): A fluorescent (fluorescein isothiocyanate (FITC)) substrate that when treated with proteases releases FITC that has an excitation at 485nm and emission at 535nm. Supplied lyophilized.
Simply incubate your sample in the reagent provided and obtain results. The kit uses dye-labeled protein conjugate as protease substrate, which allows nanogram level detection. The absorbance of dye-labeled peptide is measured at 574nm for determination of protease activity. The kit is sufficient for 50 assays in a micro well format.
Información para pedidos: Accessories not included.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty