Single Tissue Multiple Species Blot, G-Biosciences
Supplier: G-Biosciences
TB30 TB32 TB33 TB35 TB28 TB29 TB31 TB34 TB36
82023-188EA 705.7 USD InStock
82023-188 82023-192 82023-194 82023-198 82023-184 82023-186 82023-190 82023-196 82023-200
Single Tissue Multiple Species Blot, G-Biosciences
Blotting Kits
G-Biosciences offers a selection of ready-to-screen, single tissue, multi-species blots (human, mouse and rat).
G-Biosciences' premade blots are prepared with the appropriate GenLysate™ and 50 μg protein is loaded onto a 4-20% denaturing polyacrylamide gel, along with a prestained molecular weight marker. After the proteins are resolved, they are transferred to a PVDF membrane, using our Efficient Western Transfer Buffer.
G-Biosciences' premade blots are ready to be blocked and probed with the antibodies of choice. Note: We recommend that customers contact our technical department forMore Product Information
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty