ESD Stacking Box, MFG
Supplier: Molded Fiber Glass Tray
8480005167
89022-942EA 127.9 USD InStock
89022-942
ESD Stacking Box, MFG
Boxes ESD-Safe Boxes
MFG Tray ESD stacking boxes provide optimal product protection with exceptional weight-carrying capability
Boxes possess superior physical properties over thermoplastic and corrugated products. They will not bend or sag under heavy loads and can be readily integrated into automated applications. ESD stacking boxes ensure optimum space utilization during assembly operations, and may be used to form mobile work stations. Fiberstat stacking boxes ensure complete protection from electrostatic discharge for the life of the product. They are resistant to cutting oils, greases, solvents, mildMore Product Information
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty