Condenser, Cold Finger, for Pilot Plant Reactors, Ace Glass Incorporated
Supplier: Ace Glass
5958-99
89053-856EA 203.97 USD InStock
89053-856
Condenser, Cold Finger, for Pilot Plant Reactors, Ace Glass Incorporated
Condensers
This Cold Finger Condenser is used with pilot plant reactors
Diameter of tube is 14mm. Febricated from borosilicate glass. Use with #15 Ace-Thred bushing and glass adapter to fit top 45/50 standard taper joint in 89053-810 condenser. Jacket length is measured from the lower shoulder of the bulb to the bottom of the finger. Use with 5/16-inch or 3/8-inch I.D. tubing, size C hose connections.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty