Electromagnetic Coil, Plunger Principle, Ace Glass
Supplier: Ace Glass
6590-10
89055-676EA 540.99 USD InStock
89055-676
Electromagnetic Coil, Plunger Principle, Ace Glass
Magnets
This coil is designed to operate automatic distilling heads which operate on the plunger principle, whereby the glass rod is pulled up through the center of the magnet.
For operation at 120 volts, 50/60 Hz AC maximum, or at 24 volts DC maximum
For intermittent service only; 30 seconds maximum ON, 25% maximum duty cycle.
Diameter of the hole is 12.7 mm; Diameter of the coil is 38 mm; height is 32 mm.
Información para pedidos: It is supplied complete with necessary wiring to connect into the system.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty