Three-Way High Vacuum Stopcock Easy Action Plug with Tef-Cap O-Ring, Ace Glass
Supplier: Ace Glass
8190-203 8190-205
89062-416EA 383.11 USD InStock
89062-416 89062-418
Three-Way High Vacuum Stopcock Easy Action Plug with Tef-Cap O-Ring, Ace Glass
Stopcocks
High vacuum three-way stopcock for use in combining materials in proportion or as a common feed distributed in proportion
With Tef-Cap O-Ring seals that eliminate exposure to corrosive materials. For use to 3.5 Kg/cm2 internal pressure. With variable openings from 0–3 mm to 0–5 mm. An ultimate vacuum of 10-7 can be realized with the standard three seals. Smooth acting valve permits fine adjustment of opening. Front ring seal makes a positive closure against a precision formed heavy glass seat. Hooded handle is permanently attached to PTFE plug and is therefore not bakeable. O-Rings areMore Product Information
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty