Air Stirrer, Heavy Duty, High Torque, Ace Glass
Supplier: Ace Glass
13370-10
89077-664EA 934.23 USD InStock
89077-664
Air Stirrer, Heavy Duty, High Torque, Ace Glass
Stirrers Overhead Stirrers
The compact, heavy duty air motor features a highly damped muffler
Complete unit consists of an air hose with a snap coupling on motor end, and a shut-off valve on the other end. Motor will develop 1½hp at 90lbs. of air pressure. Speeds are variable from 300rpm to 3000rpm. The motor consumes approximately 42cfm at 3000rpm. The shaft measures 12.7mm (½") in diameter.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty