Corning® Pipette Tips, Universal, Corning
Supplier: Corning
4844 4845 4862 4863 4864 4865 4866 4867 4868 9032 4806 4803 4804 4860 4713 4715 4710 4846
29442-654CS 434.71 USD InStock
29442-654 29442-656 29442-680 29442-682 29442-684 29442-686 29442-688 29442-690 29442-692 29442-706 29443-270 29443-348 29443-350 29443-352 89089-706 89089-956 89089-958 29442-658
Corning® Pipette Tips, Universal, Corning
Pipette Tips
Corning® universal fit tips are designed to provide a reliable fit with all major brand pipettors.
  • Beveled orifice helps ensure accurate fluid delivery
  • All tips are certified RNase-/DNase-free and non pyrogenic
  • Stack rack tips feature a stack of five racks, each containing 96 tips, for a total of 480 tips in a space-saving design
  • Bulk packed tips are non sterile and very economical
1 to 200 µl universal fit tips are graduated at the 10 µl, 50 µl, and 100 µl volumes.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty