VWR® TraceClean® Boston Round Glass Jars, Narrow Mouth, Clear
Supplier: VWR International
89126-548 89126-550 89126-552 89126-562 89126-656
89126-548CS 77.82 USD InStock
89126-548 89126-550 89126-552 89126-562 89126-656
VWR® TraceClean® Boston Round Glass Jars, Narrow Mouth, Clear
Bottles Glass Bottles
TraceClean® glass jars are used primarily for the collection of aqueous media for testing semi-volatiles, pesticides, PCBs, diesel range organics, TPH, metals, and phenolics.
Quality Assured® with VWR® Logo Sample ID Label products, a lot certificate of analysis is generated with the analytical results obtained including the concentration reporting limits and units of measurement. This certificate of analysis is included in each product case. Please refer to the applicable Certificate of Analysis for certified analytes and corresponding reporting limits.
Secondary uses includeMore Product Information
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty