BIO-PROBE® Blur 8 Probe, Enzo Life Sciences
Supplier: SAFE AND SURE NEDERLAND B.V.
ENZ-40849
89165-736EA 415.29 USD InStock
89165-736
BIO-PROBE® Blur 8 Probe, Enzo Life Sciences
Nucleic Acid Reagents Primers and Probes
The Blur 8 BioProbe® labeled probe is prepared by nick translation of a clone of the human alu repeat sequence.
Fragment size range: 100-1000 base pairs (as estimated by agarose gel electrophoresis).
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty