Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bio-16-UTP, Enzo Life Sciences

Supplier: SAFE AND SURE NEDERLAND B.V.

ENZ-42814
89165-832EA 362.97 USD InStock
89165-832
Bio-16-UTP, Enzo Life Sciences
Nucleic Acid Reagents Nucleotides
Bio-16-UTP (Biotin-16-uridine-5’-triphosphate) can replace UTP for in vitro transcription reaction catalysed by T3, T7 or SP6 RNA polymerases.

  • Appearance: Clear, colorless liquid
  • Purity: ≥93% (HPLC)
  • Concentration: 10 mM
  • Extinction coefficient: 10,700 M-1 cm-1 (240 nm, pH 7,0)

The biotin-labelled RNA transcripts produced by these reactions are suitable for a wide range of applications such as nucleic acid hybridisation, sequencing, and genome analysis.

The biotin-labelled hybridised probes can be detected by a reporter molecule linked to streptavidin, avidin, or anit-biotin antibody. Such a complex can be detected directly, e.g. by excitation of a fluorophore conjugated to streptavidin, or indirectly, e.g. using an enzyme conjugate that can produce an insoluble coloured precipitate.


Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty