VWR® Circulating Open Bath Systems
Supplier: VWR International
89202-890 89202-894 89202-898 89202-910 89202-914 89202-918
89202-890EA 1874.26 USD InStock
89202-890 89202-894 89202-898 89202-910 89202-914 89202-918
VWR® Circulating Open Bath Systems
Circulating Thermostats Open Circulator Systems Baths with Detachable Circulators
VWR® Circulating Open Bath Systems include MX Temperature Controller and choice of stainless steel or polycarbonate reservoir.
  • Chemical-resistant
  • Polycarbonate or stainless steel fabrication
  • May be equipped with lidded openings for optional cooling coils
The DuraTop™ chemical-resistant bridge and bath cover are easy to clean and stay cooler at higher bath temperatures.
The transparent polycarbonate reservoir lets you keep samples in clear view. Elevated tank bottom provides secure handholds when lifting or relocating bath and is stable on unevenMore Product Information
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
PRODUCTOS COMPLEMENTARIOS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty