SGE Syringes, eVol® XR Digital Analytical Syringe, Trajan Scientific and Medical
Supplier: Trajan Scientific and Medical
2910020 2910022 2910024 2910021 2910023 2910025 2910026 2910027 2910320 2910322 2910324 2910035 2910037 2910335 2910329 2910029
89194-536EA 2261.55 USD InStock
89194-536 97049-424 97049-426 97049-428 89205-470 89205-472 89205-474 89205-476 89205-478 89205-480 89205-482 89205-484 89236-486 89236-488 89236-490 89236-494 89368-640 89368-642
SGE Syringes, eVol® XR Digital Analytical Syringe, Trajan Scientific and Medical
Syringes Syringes for Chromatography
eVol® XR is the coupling of two precision devices: a digitally controlled electronic drive and an XCHANGE® enabled analytical syringe.
  • Ergonomic, comfortable and easy to use
  • Variable speed of operation
  • Easily and quickly changed to dedicate to individual liquids or methods
  • Easily calibrated by operators
  • Inject directly onto a chromatography column with a consistent flow rate
The device is easily calibrated by operators, and calibration factors saved for each syringe, enabling laboratories to comply with stringent global laboratory standards (e.g. GLP,More Product Information
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty