Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-SLC12A1 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX47166
89270-286EA 640.14 USD InStock
89270-286
Anti-SLC12A1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to SLC12A1 (N-terminal)

Recommended Dilutions: For Western Blot: Use at 1.0 ug/ml. For IHC-P: Use at 4-8 ug/mL.

Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.


Type: Primary
Antigen: SLC12A1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Dog, Rat

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty