Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-RBM11 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX47213
89270-354EA 624.43 USD InStock
89270-354
Anti-RBM11 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to RBM11 (Middle)

Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: RBM11
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty