Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CTSA Rabbit Polyclonal Antibody

Supplier: Genetex
GTX86895
89289-364EA 629.01 USD InStock
89289-364
Anti-CTSA Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Cathepsin A (cleaved Arg326)

We recommend the following starting dilutions: WB: 1:500~1:1000 ELISA: 1:10000Optimal dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CTSA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty