Amplitube™ PCR Reaction Tubes, 0.2 ml, Simport Scientific
Supplier: Simport Scientific
T325-1N T325-1G T325-1R T325-1B T325-1Y T325-2B T325-2G T325-2N T325-2R T325-2Y T325-2VB T325-2VG T325-2VR T325-1VN T325-1VB T325-1VG T325-1VR T325-1VY T325-2VN T325-2VY
60872-628CS 65.19 USD InStock
60872-628 60872-632 60872-636 60872-630 60872-638 89429-014 89429-016 89429-018 89429-020 89429-022 10030-906 10030-908 10030-910 10030-912 10030-914 10030-916 10030-934 10030-936 10030-938 10030-940
Amplitube™ PCR Reaction Tubes, 0.2 ml, Simport Scientific
Tubes PCR Tubes
Amplitube™ PCR Reaction tubes are made of transparent superior grade polypropylene for improved viewing of the sample contents.
  • Attached hinged caps are either dome or flat-topped
  • Caps provide positive sealing during thermal cycling and prevent evaporation
  • Caps can be easily opened and closed with one hand
  • Autoclavable
Tubes are available with or without an integral shield that prevents contamination with surface of lid. A frosted writing surface is provided for sample identification.
Ultra-thin wall design ensures rapid thermal transfer andMore Product Information
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty