Storage Canes for Cryovial® Tubes, Simport
Supplier: Simport Scientific
T313
89511-292CS 85.3 USD InStock
89511-292
Storage Canes for Cryovial® Tubes, Simport
Cryogenic Canes
These 290 mm long storage canes for Cryovial® tubes are made of aluminium and can be used for storage of up to five 1.2 or 2 mL tubes in liquid nitrogen containers.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty