BD Taxo™ VX Factor Discs, BD Diagnostic Systems
Supplier: Seca Med.Waagen u.Meßsysteme
231731
90002-556PK 135.17 USD InStock
90002-556
BD Taxo™ VX Factor Discs, BD Diagnostic Systems
Microbiology Test Systems Microorganism Tests
BD Taxo™ X Factor Discs are used for differentiating Haemophilus species.
These paper discs are round, white 1/4” with “v” printed on both sides.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty