BD BBL™ Brand BD Crystal™ Identification Kits, BD Diagnostics
Supplier: CYTYC UK LTD
245250 245000 245240
90003-360EA 431.99 USD InStock
90003-360 90003-366 90003-380 90003-356 BB4345041 90003-378
BD BBL™ Brand BD Crystal™ Identification Kits, BD Diagnostics
Stains Microbiology Stains
These kits feature a miniaturized, easy-to-use identification method employing modified conventional, fluorogenic, and chromogenic substrates
Following an incubation period, the sample can be manually read on the Crystal panel viewer to obtain a profile number. Alternatively, the panel may be read in the Crystal AutoReader. The AutoReader, in conjunction with Crystal MIND software, automatically reads and interprets panel results.
Kits are available to identify a wide range of gram-negative, gram-positive, and anaerobic organisms. Kits provide one-step convenience, easy inoculation, and a comprehensive database (list of organisms that can be identified).
Información para pedidos: Each kit contains supplies for 20 identifications.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty