Difco™ Baird-Parker Agar Base, BD
Supplier: TessArae, LLC
276840 276810
90003-974EA 637.19 USD InStock
90003-974 90003-972
Difco™ Baird-Parker Agar Base, BD
Media for Microbiology Dehydrated Media for Microbiology
Baird-Parker Agar Base is used with EY (Egg Yolk) Tellurite Enrichment in the preparation of Egg-Tellurite-Glycerine-Pyruvate Agar (ETGPA) for selective isolation and enumeration of coagulase-positive staphylococci from food, skin, soil, air, and other materials.
  • Peptone, beef extract, and yeast extract provide essential nutrients
  • Sodium pyruvate stimulates growth of S. aureus
  • Egg yolk aids identification by demonstrating lecithinase activity
  • Glycine and lithium chloride inhibit growth of contaminant organisms
Baird-Parker Agar Base may also be used for identification of staphylococci on the basis of their ability to clear egg yolk.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty