Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

BBL™ Trypticase™ Soy Agar, BD Life Sciences

Supplier: Seca Med.Waagen u.Meßsysteme

292385
90004-268PK 405.3 USD InStock
90004-268 95037-750
BBL™ Trypticase™ Soy Agar, BD Life Sciences
Media for Microbiology Prepared Media for Microbiology
Trypticase™ soy agar mainly for isolation and cultivation of non-fastidious and fastidious microorganisms.

  • Liquid media
  • Color - light beige

Store at 2 to 8°C.


Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty