BBL™ MacConkey II Agar/Levine EMB Agar, BD Biociences
Supplier: CYTYC UK LTD
295969
90008-404PK 209.48 USD InStock
90008-404
BBL™ MacConkey II Agar/Levine EMB Agar, BD Biociences
Media for Microbiology Dehydrated Media for Microbiology
MacConkey II agar is a slightly selective and differential medium for the detection of coliform organisms and enteric pathogens.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty