VWR® Premium Frozen Section Compound
Supplier: VWR International
95057-838
95057-838CS 330.51 USD InStock
95057-838
VWR® Premium Frozen Section Compound
Stains Microscopy and staining reagents
Frozen section compound is a fast-acting, water-soluble embedding compound designed for use in frozen sectioning.
  • Water-soluble embedding compound
  • Clear siding for quick inspection
  • Minimal section curling
The compound bonds and encapsulates tissue specimens to the object holder for cryosectioning. At a working temperature of -20°C (-4°F), it provides optimal sectioning consistency with minimal section curling. Each bottle features an extended length tip for increased accuracy in dispensing. Highly recommended for surgical pathology laboratories. Available in a clear format for better visualization of small specimens. Size: 118 mL (4 oz.).
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty