Bulk Syringe Caps, Non Sterile, Air-Tite Products
Supplier: Air-Tite
BUCC
97001-202CS 42.91 USD InStock
97001-202
Bulk Syringe Caps, Non Sterile, Air-Tite Products
Closures Test/Sample Tube Closures
These bulk syringe caps are very cost-efficient.
  • Fits all Luer lock and luer slip syringes
  • Bulk orders
  • Clear
For capping off and sealing syringes, these clear polypropylene syringe caps fit all Luer slip and Luer lock syringes.
Envase: Packed in bulk and non-sterile.
Order Now
ORDER
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
Collapse All
Description Type For use with Packaging Supplier No. VWR Catalog Number Unit Price Quantity
Bulk Syringe Caps Non sterile Laboratory and Veterinary Use Only 1000/box BUCC 97001-202 Case of 1,000
Retrieving
Restricted
restricted This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty