InLab® Reference Pro Electrode, Sleeve Junction, METTLER TOLEDO®
Supplier: Mettler Toledo
51343191
97001-678EA 454.71 USD InStock
97001-678
InLab® Reference Pro Electrode, Sleeve Junction, METTLER TOLEDO®
Electrodes pH Electrodes
Electrode is ideal for measurements in ion-selective half-cells
  • Moveable PTFE sleeve junction
  • Bridge gel electrolyte chamber
  • S7 screw head connector
Features moveable PTFE sleeve junction and bridge gel electrolyte chamber. S7 screw head connector is compatible with various cables, so electrode may be used with a wide range of meters.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty