Temperature Chart Recorders, Dickson®
Supplier: Dickson
C015 C017 C181 C070 C010 C012 C083 C205 C206 C207 C208 SL4100 SL4350 C210
12444-170PK 118.62 USD InStock
12444-170 12444-172 61280-105 61280-128 61280-160 61280-206 61280-208 61280-235 61280-237 97006-892 97006-894 97006-896 97006-898 97006-900 97006-908 97006-910 97006-902
Temperature Chart Recorders, Dickson®
Chart Recorders Recorders
Recorders are ideal for monitoring long-term temperature trends in areas where proper temperatures are critical to product stability.
  • Cord-free, portable, and easy to use
  • Recorders can be placed directly in an environment that needs to be monitored, and mounted on a desk or wall
  • Feature spiral-wound bimetallic strip transducer, hinged door, built-in slot for optional tie-down cable and lock, battery door
  • Durable, corrosion-resistant black ABS and polycarbonate enclosure
  • Recorders come with or without digital LCD, and can be set to 7-day or 24-hour
More Product Information
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
SERVICIOS
Servicios: VWRCATALYST™ offers validation services including IQ, OQ, and PQ protocol development and execution, as well as asset management and calibration services (US only); call 888-793-2300 for more details.
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty