GFAAS Mixed Standard, Matrix per Volume 5% HNO₃, PerkinElmer
Supplier: PERKINELMER U.S. LLC
N9300244
97011-978EA 566.43 USD InStock
97011-978
GFAAS Mixed Standard, Matrix per Volume 5% HNO₃, PerkinElmer
Standards Cationic Standards Multi-Element Standards
The mixed standard, matrix per volume 5% HNO₃, concentration of elements are 5 mg/l Beryllium and Cadmium, 10 mg/l Silver, 20 mg/l Chromium, Iron and Magnesium, 50 mg/l Barium, Cobalt, Copper and Nickel, 100 mg/l Aluminium, Arsenic, Lead, Antimony, Selenium and Thallium.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty