Gallium (Ga) Lumina™ Hollow Cathode Lamp, PerkinElmer
Supplier: PERKINELMER U.S. LLC
N3050128
97013-332EA 7369.81 USD InStock
97013-332
Gallium (Ga) Lumina™ Hollow Cathode Lamp, PerkinElmer
Lights Hollow Cathode Lamps
Single Element Lumina™ hollow cathode lamp (HCL) for the detection of elemental Gallium (Ga). Series N30501XX Lumina™ 50 mm (2") diameter single-element lamps are designed to be used with PinAAcle™ and AAnalyst™ atomic absorption spectrometer series of instruments.
Lamp is designed for use with and tested on spectrometers to assure compatibility and the highest performance. Offers a wide range of single-element and multi-element HCLs, which are ideal for determining most elements by atomic absorption spectroscopy. To prolong the life of a Lumina™ hollow cathode lamp (HCL), produce lampsMore Product Information
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty