RS-P28 Printer, METTLER TOLEDO®
Supplier: Mettler Toledo
11124324
97016-772EA 1904.28 USD InStock
97016-772
RS-P28 Printer, METTLER TOLEDO®
Printers Document Printers
The printer supports the connected instrument languages, whether it operates in English, Russian, Chinese (balance), Japanese or Latin charaters.
  • Statistical and totaling applications
  • Fulfills GxP requirements
  • RS232 interfaces
  • Dot matrix printing technology
Build in real time and date allows users to record all measurements with customized data printouts.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty