Plastic Sample Bottle
Supplier: Mettler Toledo
51300240
97025-902EA 38.93 USD InStock
97025-902
Plastic Sample Bottle
Electrochemistry Meters pH and Multi-Parameter Meters
Plastic sample bottle, an useful meter accessories for uGo carrying case.
Compatible meter: FiveEasy, FiveEasy Plus, FiveGo, Seven2Go, Seven2Go Pro, SevenCompact, SevenExcellence, SevenGo Duo Pro, SevenGoDuo
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty