MicroSolv™ Pharma Grade Vials, Caps and Convenience Kits
Supplier: MicroSolv Technology Corp
9502C-0-0CV 9502C-0CV 9502C-0DCV 9502C-1CP 9502C-1WAP 9502C-1WCP 9502C-2CP 9502C-2WAP 9502C-2WCP 9502C-3WAP 9502C-3WCP 9502C-4CP 9502C-4WAP 9502C-4WCP 9502C-WAV 9502C-WCV 9502C-WDCV 9502S-06-0CP 9502S-06-0DCP 9502S-06-SWAP 9502S-06-WAP 9502S-06-WCP 9502S-0CP 9502S-0CV 9502S-0DCP 9502S-0DCV 9502S-0WAP 9502S-0WCP 9502S-1CP 9502S-1DCP 9502S-1WAP 9502S-1WCP 9502S-2CP 9502S-2WAP 9502S-2WCP 9502S-3CP 9502S-3DCP 9502S-3WAP 9502S-3WCP 9502S-4CP 9502S-4WAP 9502S-4WCP 9502S-WAV 9502S-WCV 9502C-5CP 9502C-5WAP 9502C-5WCP 9504S-0AV 9504S-0CV 9504S-WAV 9504S-WCV 9502S-WDCV 9502C-WDAV 9504S-8CP 9502S-E-35-0PB 9504S-2CP 9504S-2WAP 9504S-2WCP 9504S-3CP 9504S-3WAP 9504S-3WCP 9504S-1CP 9504S-1WAP 9504S-1WCP 9502S-E-3DAP
97052-286PK 42.64 USD InStock
97052-286 97052-292 97052-294 97052-328 97052-330 97052-332 97052-344 97052-346 97052-348 97052-358 97052-360 97052-382 97052-384 97052-386 97052-426 97052-428 97052-430 97052-464 97052-466 97052-470 97052-472 97052-474 97052-486 97052-488 97052-490 97052-492 97052-494 97052-496 97052-508 97052-510 97052-512 97052-514 97052-522 97052-524 97052-526 97052-538 97052-540 97052-542 97052-544 97052-552 97052-554 97052-556 97052-594 97052-598 97055-290 97055-292 97055-294 97055-366 97055-368 97055-388 97055-390 10040-092 10804-168 10804-198 10804-668 10811-024 10811-026 10811-028 10811-030 10811-032 10811-034 10811-080 10811-082 10811-084 76436-422
MicroSolv™ Pharma Grade Vials, Caps and Convenience Kits
Vials
MicroSolv™ brand family of chromatography N51A pharma grade clear or amber glass vials, with or without a write on patch.
  • Pharmaceutical grade specifications chosen by QC and R&D departments when reliability is desired
  • Made with first Hydrolytic, N51A borosilicate glass
Glass and plastic specifications that meet or exceed demanding laboratory requirements and have minimal impact on sensitive samples. Vials that are produced using computerized cameras as they are tooled to assure perfect fit and form.
MicroSolv™ Brand vials and caps wereMore Product Information
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty