Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Uridine for biochemistry

Supplier: VWR Solon Production

0975-50G
97062-542CS 2164.56 USD InStock
97062-542
Uridine for biochemistry
Uridine


Formula: C₉H₁₂N₂O₆
MW: 244.20 g/mol
Melting Pt: 166 °C…167 °C
Storage Temperature: Ambient
MDL Number: MFCD00006526
CAS Number: 58-96-8
EINECS: 200-407-5
Order Now
Specification Test Results
Em (261nm, 0.15M Phosphate Buffer, pH 7.3) >= 10100

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty