Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Alizarin red S (sodium salt) for biochemistry

Supplier: VWR Solon Production
Warning

9436-25G
97062-616EA 92.08 USD InStock
97062-616
Alizarin red S (sodium salt) for biochemistry
Alizarin Red S (sodium salt)


Formula: C₁₄H₇NaO₇S
MW: 342.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013049
CAS Number: 130-22-3
EINECS: 204-981-8
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty