VWR® Malt Extract
Supplier: VWR Solon Production
J873-500G
97063-426CS 246.24 USD InStock
97063-426 97063-428 97063-430
VWR® Malt Extract
Media for Microbiology Dehydrated Media for Microbiology
VWR® offers high purity extracts used for media supplements.
  • Used for the preparation of media for the detection of yeasts and molds. Hazards, N/A. Bacteriological Grade
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty