Brilliant II SYBR® MM with ROX, Agilent Technologies
Supplier: C + P Möbelsysteme GmbH & Co.KG
600835 600830
97066-494EA 777.78 USD InStock
97066-494 97066-502 97066-496 97066-504
Brilliant II SYBR® MM with ROX, Agilent Technologies
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
Brilliant II SYBR® QPCR and QRT-PCR master mix kits with high/low Rox offer superior sensitivity for improved quantification and reproducibility.
  • Improved performance with earlier Ct detection across a wide dynamic range
  • Increased sensitivity of detection to low copy numbers
It also shows more consistent performance across a wide range of template concentrations and different targets.
Precaución: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty