Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cetrimonium chloride ≥96%

Supplier: Origen Biomedical, Inc
Danger

041861-06 041861-14 041861-22 041861-36
AA41861-06EA 38.8 USD InStock
AA41861-06 AA41861-14 AA41861-22 AA41861-36
Cetrimonium chloride ≥96%
Cetrimonium chloride
MDL: MFCD00011773 Beilstein Registry No.: 3657974

Purity: 96% CAS No.: 112-02-7 Molecular Formula: C19H42ClN Formula Weight: 320.00 Physical Form: Powder Density: 0.968 Appearance: Off-white Odor: Odorless Melting Point: 234° Beilstein Reference: 3657974 UN No.: 1759 MDL No.: MFCD00011773


Formula: C₁₉H₄₂ClN
MW: 320 g/mol
Melting Pt: 234 °C
Density: 0.968
Storage Temperature: Ambient
MDL Number: MFCD00011773
CAS Number: 112-02-7
EINECS: 203-928-6
UN: 2923
ADR: 8 III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty