Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-1-pentanol 95%

Supplier: Origen Biomedical, Inc
Warning

L12088-09 L12088-18
AAAL12088-09EA 93.2 USD InStock
AAAL12088-09 AAAL12088-18
5-Chloro-1-pentanol 95%
5-Chloro-1-pentanol
Purity: 95% CAS No.: 5259-98-3 Molecular Formula: Cl(CH2)5OH Formula Weight: 122.60 Refractive Index: 1.4540 Boiling Point: 102-103°/8mm MDL No.: MFCD00039556


Formula: C₅H₁₁ClO
MW: 122.59 g/mol
Boiling Pt: 102…103 °C (8 mmHg)
Density: 1.060
Storage Temperature: Ambient
MDL Number: MFCD00039556
CAS Number: 5259-98-3
EINECS: 226-067-8
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty