Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Iodobenzyl alcohol 97%

Supplier: Origen Biomedical, Inc
Warning

L19278-06 L19278-03
AAAL19278-06EA 123 USD InStock
AAAL19278-06 AAAL19278-03
4-Iodobenzyl alcohol 97%
4-Iodobenzyl alcohol
Purity: 97% CAS No.: 18282-51-4 Molecular Formula: C7H7IO Formula Weight: 234.04 Melting Point: 72-74° Beilstein Reference: 1931621


Formula: C₇H₇IO
MW: 234.04 g/mol
Melting Pt: 72…74 °C
Storage Temperature: Ambient
MDL Number: MFCD01732720
CAS Number: 18282-51-4
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty