Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl glycolate 94%

Supplier: Origen Biomedical, Inc
Warning

H35711-03
AAH35711-03EA 580 USD InStock
AAH35711-03 AAH35711-MD
tert-Butyl glycolate 94%
tert-Butyl glycolate
tert-Butyl glycolate, 95%

Purity: 94% CAS No.: 50595-15-8 Molecular Formula: C6H12O3 Formula Weight: 132.16 Storage Temperature: 2°C to 8°C Physical Form: Low melting solid Appearance: Colorless to yellow MDL No.: MFCD03452739


Formula: C₆H₁₂O₃
MW: 132.159 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03452739
CAS Number: 50595-15-8
Order Now
Specification Test Results
Assay 95%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty