CrossLab Ultra Inert Liners for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
5190-2294 5190-2295 5190-3163 5190-3164 5190-2293 5190-3165 5190-3167 5190-3169 5190-3171 5190-3983 5190-4007 5190-4047 5190-4048 5190-6168 5190-2292 5190-3162 5190-3166 5190-3168 5190-3172 5190-4056 5190-5105-005 5190-5112 5190-5112-005 5190-7020 5190-3170 5190-3173 5190-5105 5190-7011 5190-5105-025 5190-7012 5190-7013 5190-7014 5190-7045
AG5190-2294EA 49.8 USD InStock
AG5190-2294 AG5190-2295 AG5190-3163 AG5190-3164 AG5190-2293 AG5190-3165 AG5190-3167 AG5190-3169 AG5190-3171 AG5190-3983 AG5190-4007 AG5190-4047 AG5190-4048 AG5190-6168 AG5190-2292 AG5190-3162 AG5190-3166 AG5190-3168 AG5190-3172 AG5190-4056 AG5190-5105005 AG5190-5112 AG5190-5112005 AG5190-7020 AG5190-3170 AG5190-3173 AG5190-5105 AG5190-7011 AG51905105-025 AG5190-7012 AG5190-7013 AG5190-7014 AG5190-7045
CrossLab Ultra Inert Liners for GC, Agilent Technologies
Chromatography Supplies Chromatography Inlet Liners
Agilent Ultra Inert inlet liners prevent adsorption and ensures accurate sample transfer onto the GC column.
  • Glass-fritted liners now available for splitless and universal applications to help increase reproducibility and extend liner life
  • Consistent high level of inertness for greater sensitivity, accuracy, and reproducibility
  • Ultra Inert split and splitless liners, with and without wool, for a wide variety of applications
  • Liners with inert wool retain nonvolatiles, making trace-level analysis of difficult active compounds easier
  • Unique touchless
More Product Information
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty