Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Propanol ≥70%, Production Grade, VWR Chemicals BDH®

Supplier: VWR International
Danger

BDH2014-1GLP BDH2014-55GLP BDH2014-5GLP BDH2014-5GLMS
BDH2014-1GLPCS 626.29 USD InStock
BDH2014-1GLP BDH2014-55GLP BDH2014-5GLP BDH2014-5GLMS
2-Propanol ≥70%, Production Grade, VWR Chemicals BDH®
2-Propanol
Clear, colorless liquid.


Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 82
Melting Pt: -89
Density: 0.8…0.92 g/cm³ (20 °C)
Flash Pt: 12
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1987
ADR: 3 II
Merck Index: 13,05228
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=77f11bcf690db67b8cf7c1dbbc5d3a4d2586aeed&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Clear, Colorless
Specific Gravity at 60°F (0.872 – 0.883) g/mL

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty