Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ammonia solution ≥25% (NH₃ basis) in water, Puriss. p.a. plus analytical reagent, Fluka™

Supplier: BURDICK & JACKSON FKA SIGMA
Danger

17093-1L
BJ17093-1LEA 121.78 USD InStock
BJ17093-1L
Ammonia solution ≥25% (NH₃ basis) in water, Puriss. p.a. plus analytical reagent, Fluka™
Ammonia solution
  • Exceeds standard ACS specifications with additional trace metals analysis
  • Products with a guarantee certificate and/or suitable for the stated analytical application.


Formula: NH₃
MW: 35.05 g/mol
Boiling Pt: 37.7 °C (1.013 hPa)
Melting Pt: –92 °C
Density: 0.90 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
UN: 2672
ADR: 8 III
Order Now
Specification Test Results
Assay Min. 25 %
Non-volatile matter Max. 10 ppm
Silver (Ag) Max. 0.01 ppm
Aluminium (Al) Max. 0.01 ppm
Arsenic, antimony (as As) Max. 0.01 ppm
Gold (Au) Max. 0.01 ppm
Boron (B) Max. 0.01 ppm
Barium (Ba) Max. 0.01 ppm
Beryllium (Be) Max. 0.01 ppm
Bismuth (Bi) Max. 0.01 ppm
Calcium (Ca) Max. 0.01 ppm
Cadmium (Cd) Max. 0.01 ppm
Cobalt (Co) Max. 0.01 ppm
Chromium (Cr) Max. 0.01 ppm
Cesium (Cs) Max. 0.01 ppm
Copper (Cu) Max. 0.01 ppm
Iron (Fe) Max. 0.01 ppm
Gallium (Ga) Max. 0.01 ppm
Germanium (Ge) Max. 0.01 ppm
Indium (In) Max. 0.01 ppm
Potassium (K) Max. 0.01 ppm
Lithium (Li) Max. 0.01 ppm
Magnesium (Mg) Max. 0.01 ppm
Manganese (Mn) Max. 0.01 ppm
Molybdenum (Mo) Max. 0.01 ppm
Sodium (Na) Max. 0.01 ppm
Nickel (Ni) Max. 0.01 ppm
Lead (Pb) Max. 0.01 ppm
Platinum (Pt) Max. 0.01 ppm
Tin (Sn) Max. 0.01 ppm
Strontium (Sr) Max. 0.01 ppm
Titanium (Ti) Max. 0.01 ppm
Thallium (Tl) Max. 0.01 ppm
Vanadium (V) Max. 0.01 ppm
Zinc (Zn) Max. 0.01 ppm
Zirconium (Zr) Max. 0.01 ppm
Explosion Limit 27 %
Chloride (Cl) Max. 0.2 ppm
Phosphate (PO₄) Max. 0.2 ppm
Sulfate (SO₄) Max. 1 ppm

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty