Biomek® Pipette Tips for Biomek Liquid Handlers, Beckman Coulter®
Supplier: Beckman Coulter
B01122 379501 394263 719013 987935 987936 A21578 A21582 A22287 B01114 B01123 140505 394622 394627 719010 719223 B01089 B01091 B01101 B01102 B01113 379504 A22288 B01112 379505 379506 A21586 B01090 B01103 717256 B01124 373687 379502 379503 717251 717252 717253 717254 717255 719011 719012 719222 987925 B85955 B85965 B85981 B85739 B85745 B85753 B85756 B85749 B85760 B85877 B85903 B85945 B85961 B85975 B85936 B85881 B85764 B85884 B85767 B85906 B85926 B85929 B85771 B85888 B85911 B85775 B85915 B85892 B85872 B85917 B85894 B85940 B85959 B85971 B85922 B85899 C41861 C41862 C41863 C41864 C41865
BKB01122CS 264.95 USD InStock
BKB01122 BK379501 BK394263 BK719013 BK987935 BK987936 BKA21578 BKA21582 BKA22287 BKB01114 BKB01123 BK140505 BK394622 BK394627 BK719010 BK719223 BKB01089 BKB01091 BKB01101 BKB01102 BKB01113 BK379504 BKA22288 BKB01112 BK379505 BK379506 BKA21586 BKB01090 BKB01103 BK717256 BKB01124 BK373687 BK379502 BK379503 BK717251 BK717252 BK717253 BK717254 BK717255 BK719011 BK719012 BK719222 BK987925 76111-652 76111-654 76111-656 76111-658 76111-660 76111-662 76111-664 76111-666 76111-668 76111-864 76111-882 76111-890 76111-892 76111-894 76111-896 76112-010 76112-012 76112-014 76112-016 76112-018 76112-020 76112-022 76112-024 76112-026 76112-028 76112-030 76112-032 76112-034 76112-036 76112-038 76112-040 76112-042 76112-044 76112-046 76112-048 76112-050 76334-646 76334-936 76334-938 76334-940 76334-942
Biomek® Pipette Tips for Biomek Liquid Handlers, Beckman Coulter®
Pipette Tips
Biomek® Pipette Tips from Beckman Coulter® are the only tips on the market that are designed, validated, and approved for use on Biomek® Liquid Handlers.
  • Color-coded racks
  • Easy visual checks
Tip definitions, pipetting techniques, and templates are built into Biomek® software. For best in total system performance, use only Beckman Coulter® validated pipette tips on Biomek® workstations.
Products that are designated as Sterile are either sterilized using Gamma radiation or ethylene oxide (EtO) gas in accordance with ANSI/AAMI/150 11135 or ANSI/AAMI/More Product Information
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty