Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Polyethyleneimine solution, M.W. ≈ 70,000 30% (w/w) in water, branched

Supplier: BEANTOWN CHEMICAL - BTC

219710-100G 219710-25G 219710-500G
BT219710-100GEA 162.35 USD InStock
BT219710-100G BT219710-25G BT219710-500G
Polyethyleneimine solution, M.W. ≈ 70,000 30% (w/w) in water, branched
Polyethyleneimine
CAS: 9002-98-6; MDL No: MFCD00803910; RTECS: TQ7915000 Liquid; Linear Formula: (-NHCH2CH2-)x[-N(CH2CH2NH2)CH2CH2-]y

  • RTECS: TQ7915000
  • Liquid


Formula: (CH2CH2NH)n
MDL Number: MFCD00084427
CAS Number: 9002-98-6
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty