Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Barium hydroxide octahydrate, EMSURE® for analysis, Supelco®

Supplier: EMD MILLIPORE - AA – CORP
Danger

1.01737.0500
EM1.01737.0500CS 1348 USD InStock
EM1.01737.0500
Barium hydroxide octahydrate, EMSURE® for analysis, Supelco®
Barium hydroxide octahydrate


Formula: Ba(OH)₂·8H₂O
MW: 315.47 g/mol
Boiling Pt: 780
Melting Pt: 78
Density: 3.2 g/cm³ (20 °C)
Flash Pt: 21 °C (closed cup)
MDL Number: MFCD00149152
CAS Number: 12230-71-6
EINECS: 236-710-4
UN: 2923
ADR: 8 (6.1) II
Merck Index: 13,00980
Order Now
Specification Test Results
Solubility 72 g/l (20 °C)
Melting Point 78 °C
Molar Mass 315.48 g/mol
Bulk Density 900 - 1100 kg/m3
Density 2.18 g/cm3 (20 °C)
pH 12.5 (50 g/l, H2O, 20 °C) (slurry)
Hazard Class 8
Hazard Class Description Corrosive Material

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty