Extran® AP 12 Powdered Detergent for Automatic Washing, MilliporeSigma
Supplier: MilliporeSigma
1.07563.2000 1.07563.9010 1.07563.9025
EM1.07563.2000CS 438 USD InStock
EM1.07563.2000 EM1.07563.9010 EM1.07563.9025
Extran® AP 12 Powdered Detergent for Automatic Washing, MilliporeSigma
Cleaning Agents Detergents and Surface Cleansers
Active universal cleaning agent for the main wash cycle, which cleans even heavily soiled items and removes dried or burned-on residues. Particularly suitable for the removal of starch and protein residues. Extran® AP 12 alkaline powder does not foam even during heavy agitation of the solution in a washing machine.
  • Free from organic surfactants and emulsifiers
  • Powder form
  • Alkaline
  • Surfactant-free and chlorine-free
  • Free from odorants/dyestuffs
Dosing
The ideal dosing depends on the hardness of the water and the level of contamination of the item to beMore Product Information
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty