Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Palladium(II) oxide Pd 85% for synthesis, hydrogenation catalyst, Sigma-Aldrich®

Supplier: EMD MILLIPORE - AA – CORP
Danger

M-Clarity™ quality level = MQ200

8.07107.0001
EM8.07107.0001CS 817679 USD InStock
EM8.07107.0001
Palladium(II) oxide Pd 85% for synthesis, hydrogenation catalyst, Sigma-Aldrich®
Palladium(II) oxide


Formula: OPd
MW: 122.42 g/mol
Melting Pt: 870
MDL Number: MFCD00011172
CAS Number: 1314-08-5
UN: 1479
ADR: 5.1 II
Merck Index: 12,07125
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Molar Mass 122.42 g/mol
Hill Formula OPd
Chemical Formula PdO

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty