Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Taurine for synthesis, Sigma-Aldrich®

Supplier: EMD MILLIPORE - AA – CORP

M-Clarity™ quality level = MQ200

8.08616.0005 8.08616.0250 8.08616.1000
EM8.08616.0005CS 30112 USD InStock
EM8.08616.0005 EM8.08616.0250 EM8.08616.1000
Taurine for synthesis, Sigma-Aldrich®
Taurine


Formula: C₂H₇NO₃S
MW: 125.15 g/mol
Melting Pt: 300
MDL Number: MFCD00008197
CAS Number: 107-35-7
Merck Index: 13,09163
Order Now
Specification Test Results
Solubility 65 g/l (12 °C)
Melting Point 300 °C (decomposition)
Molar Mass 125.15 g/mol

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty